Anti-JNK1+JNK2+JNK3 (phospho Y185) Rabbit Monoclonal Antibody [clone: EP1597Y]
: Abcam
New Product
Rabbit monoclonal [EP1597Y] to JNK1 (phospho Y185) + JNK2 (phospho Y185) + JNK3 (phospho Y223).
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: JNK1+JNK2+JNK3
Clonality: Monoclonal
Clone: EP1597Y
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
- Catalog No:
- ABCAAB76572-100
- ABCAAB76572-1ML
- Antigen symbol:
- MAPK8/9/10
- Antigen name:
- Mitogen-activated protein kinase 8/9/10
- Antibody type:
- Primary
- Clonality:
- Monoclonal
- Conjugation:
- Unconjugated
- Clone:
- EP1597Y
- Host:
- Rabbit
- Isotype:
- IgG
- Western blot:
- Yes
- ImmunoPrecipitation:
- Yes
- Storage temperature:
- Cool
- Shipping temperature:
- Ice
Frequently Bought Together