Anti-Integrin beta 1D Mouse Monoclonal Antibody [clone: 1G2]
BIRBORB98351-0.1
: Biorbyt
- Antibody type:Primary
- Antigen name:Integrin Beta 1D
- Antigen symbol:ITGB1D
- Clonality:Monoclonal
- Clone:1G2
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Western blot:Yes
- Storage buffer:PBS containing 0,09% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:0,1 mg
Type: Primary
Antigen: Integrin beta 1D
Clonality: monoclonal
Clone: 1G2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: