Order Entry
Hungary
ContactUsLinkComponent
Anti-IgG1/IgG2a/IgG2b/IgG3 Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Anti-IgG1/IgG2a/IgG2b/IgG3 Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # BIRBORB98171-2
Supplier:  Biorbyt
Anti-IgG1/IgG2a/IgG2b/IgG3 Rabbit Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # BIRBORB98171-2
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Secondary
  • Antigen symbol:
    IgG1/IgG2a/IgG2b/IgG3
  • Clonality:
    Polyclonal
  • Conjugation:
    TRITC (Tetramethylrhodamine Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Mouse
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Size:
    2 ml
  • Pk:
    2 mL

Specifications

About this item

Type: Secondary
Antigen: Mouse IgG1 IgG2a IgG2b IgG3
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse