Anti-Ly-6G Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy7®)
Catalog # BIRBORB131007-100
Gyártó: Biorbyt
Specifikációk
- Antibody type:Primary
- Antigen symbol:LY-6G
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)/Cy7®
- Host:Rabbit
- Isotype:IgG
- Reactivity:Mouse
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Specifikációk
Termék információk
Type: Primary
Antigen: Ly-6G (Ly-6G)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse