Anti-S100A7 Rabbit Polyclonal Antibody (PE (Phycoerythrin)/Cy5®)
BIRBORB130123-100
: Biorbyt
- Antibody type:Primary
- Antigen name:S100 calcium binding protein A7
- Antigen symbol:S100A7
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)/Cy5®
- Host:Rabbit
- Isotype:IgG
- Reactivity:Rat,Mouse
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:11 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: S100A7 (S100 calcium binding protein A7)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat