Anti-Envelope Glycoprotein Yellow Fever Virus Rabbit Polyclonal Antibody (Cy5®)
BIRBORB122641-100
: Biorbyt
- Antibody type:Primary
- Antigen name:Envelope Glycoprotein Yellow Fever Virus
- Clonality:Polyclonal
- Conjugation:Cy5®
- Host:Rabbit
- Isotype:IgG
- Reactivity:Virus
- Form:liquid
- Storage buffer:10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
- Molecular weight:52 kDa
- Storage temperature:Cool
- Shipping temperature:Ice
- Immunogen:Synthetic peptide
- Purification:purified by affinity chromatography
- Size:100 µl
- Pk:100 µl
Type: Primary
Antigen: Envelope Glycoprotein Yellow Fever Virus
Clonality: polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Virus