Order Entry
Hungary
ContactUsLinkComponent
Anti-G Protein-Coupled Receptor R35/PGR10/GPR149 Rabbit polyclonal antibody
Anti-G Protein-Coupled Receptor R35/PGR10/GPR149 Rabbit polyclonal antibody
Catalog # BIRBORB109219-50
Supplier:  Biorbyt
undefined
Anti-G Protein-Coupled Receptor R35/PGR10/GPR149 Rabbit polyclonal antibody
Catalog # BIRBORB109219-50
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    G Protein-Coupled Receptor R35/PGR10/GPR149
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Immunogen:
    C-terminal domain of human
  • Purification:
    purified by immunoaffinity chromatography
  • Size:
    50 µg
  • Pk:
    50 µG

Specifications

About this item

Type: Primary
Antigen: G Protein-Coupled Receptor R35/PGR10/GPR149
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human