Order Entry
Hungary
ContactUsLinkComponent
 
Anti-HSPA5 Rabbit Polyclonal Antibody Atto Dyes and Enzyme-Protein Labels
  BIRBORB107273-100
 :  Biorbyt
undefined
Anti-HSPA5 Rabbit Polyclonal Antibody Atto Dyes and Enzyme-Protein Labels
  BIRBORB107273-100
 :  Biorbyt

 

  • Antibody type:
    Primary
  • Antigen name:
    Heat shock 70kDa protein 5
  • Antigen symbol:
    HSPA5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Storage buffer:
    PBS pH 7,4, 50% glyercol and 0,09% sodium azide
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    His-tagged rat GRP78
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Type: Primary
Antigen: HSPA5 (heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa))
Clonality: polyclonal
Clone:
Conjugation: Atto Dyes and Enzyme-Protein Labels
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat