Order Entry
Hungary
ContactUsLinkComponent
Anti-MAP-2 (2a + 2b) Mouse Monoclonal Antibody (FITC (Fluorescein))
Anti-MAP-2 (2a + 2b) Mouse Monoclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB26630-0.25
Supplier:  Biorbyt
Anti-MAP-2 (2a + 2b) Mouse Monoclonal Antibody (FITC (Fluorescein))
Catalog # BIRBORB26630-0.25
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    MAP-2A/B
  • Clonality:
    Monoclonal
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG1
  • Western blot:
    Yes
  • Storage buffer:
    0,01 M PBS, pH 7,4 containing 1% BSA.
  • Storage temperature:
    Cool
  • Concentration:
    0,1 mg/ml (OD280)
  • Shipping temperature:
    Ice
  • Purification:
    purified by Protein G chromatography
  • Size:
    250 μg
  • Pk:
    0,25 mg

Specifications

About this item

Type: Primary
Antigen: MAP-2 (2a + 2b)
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: