Order Entry
Hungary
ContactUsLinkComponent
Anti-HCV Core Antigen Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 6A1]
Anti-HCV Core Antigen Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 6A1]
Catalog # BIRBORB24126-100
Supplier:  Biorbyt
Anti-HCV Core Antigen Mouse Monoclonal Antibody (FITC (Fluorescein)) [clone: 6A1]
Catalog # BIRBORB24126-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    HCV Core Antigen
  • Antigen symbol:
    HCV
  • Clonality:
    Monoclonal
  • Clone:
    6A1
  • Conjugation:
    FITC (Fluorescein Isothiocyanate)
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG2a
  • Amino acid number:
    1 to 80
  • Storage buffer:
    PBS, pH 7,2
  • Storage temperature:
    Cool
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Recombinant protein
  • Purification:
    purified by Protein A chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Type: Primary
Antigen: HCV Core Antigen
Clonality: monoclonal
Clone: 6A1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: