Anti-HCV Core Antigen Mouse Monoclonal Antibody (Biotin) [clone: 6A1]
Catalog # BIRBORB24125-100
Supplier: Biorbyt
Specifications
- Antibody type:Primary
- Antigen name:HCV Core Antigen
- Antigen symbol:HCV
- Clonality:Monoclonal
- Clone:6A1
- Conjugation:Biotin
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG2a
- Amino acid number:1 to 80
- Storage buffer:PBS, pH 7,2
- Storage temperature:Cool
- Concentration:1 mg/ml
- Shipping temperature:Ice
- Immunogen:Recombinant Protein
- Purification:purified by Protein A chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: HCV Core Antigen
Clonality: monoclonal
Clone: 6A1
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: