Order Entry
Hungary
ContactUsLinkComponent
Anti-PiggyBac transposable element-derived protein 5 Rabbit Polyclonal Antibody
Anti-PiggyBac transposable element-derived protein 5 Rabbit Polyclonal Antibody
Catalog # BIRBORB13159-100
Supplier:  Biorbyt
undefined
Anti-PiggyBac transposable element-derived protein 5 Rabbit Polyclonal Antibody
Catalog # BIRBORB13159-100
Supplier:  Biorbyt

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    PiggyBac transposable element-derived Protein 5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Western blot:
    Yes
  • Storage buffer:
    TBS (pH 7,4), 0,5% BSA, 25% Glycerol. Preservative: 0,05% Sodium Azide
  • Storage temperature:
    Cool
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    Ice
  • Immunogen:
    Synthetic peptide
  • Purification:
    purified by affinity chromatography
  • Size:
    100 µl
  • Pk:
    100 µl

Specifications

About this item

Western Blot: 1:10000-20000; Immunofluorescence: 1:100-200

Type: Primary
Antigen: PiggyBac transposable element-derived protein 5
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: