Anti-CD45/CD14 Mouse Monoclonal Antibody (FITC (Fluorescein))/PE (Phycoerythrin)
Gyártó: Biorbyt
Tanúsítványok
Termék információk
Type: Primary
Antigen: CD45/CD14
Clonality: monoclonal
Clone:
Conjugation: FITC (Fluorescein)/PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1+IgG2a
Reactivity: Human
Specifikációk
- Catalog No:
- BIRBORB12163-20
- BIRBORB12163-50
- Antigen symbol:
- CD45/CD14
- CD45/CD14
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- FITC (Fluorescein Isothiocyanate)
- FITC (Fluorescein Isothiocyanate)
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG2a, IgG1
- IgG2a, IgG1
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Purification:
- purified by affinity chromatography
- purified by affinity chromatography
- Storage buffer:
- Phosphate-buffered solution, pH 7,4, containing 0,09% sodium azide and 0,2% (w/v) BSA
- Phosphate-buffered solution, pH 7,4, containing 0,09% sodium azide and 0,2% (w/v) BSA
- Storage temperature:
- Cool
- Cool
- Shipping temperature:
- Ice
- Ice
Specifikációk
Frequently Bought Together