Order Entry
Hungary
ContactUsLinkComponent
Anti-CLIC5 Rabbit Polyclonal Antibody
Anti-CLIC5 Rabbit Polyclonal Antibody
Catalog # ABGEAP7583A
Supplier:  Abgent
undefined
Anti-CLIC5 Rabbit Polyclonal Antibody
Catalog # ABGEAP7583A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Chlochloride Intracellular Channel 5
  • Antigen symbol:
    CLIC5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Antigen synonyms:
    hTDAG8|TDAG8
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: CLIC5 (chloride intracellular channel 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human