Anti-CNN3 Rabbit Polyclonal Antibody
Catalog # ABGEAP6581C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Nerve Growth Factor
- Antigen symbol:NGF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:HSAN5|Beta-NGF|NGFB
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: CNN3 (calponin 3, acidic)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human