Anti-PRG2 Rabbit Polyclonal Antibody
ABGEAP5732C
: Abgent
- Antibody type:Primary
- Antigen name:Phospholipid Phosphatase Related 3
- Antigen symbol:LPPR3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:middle
- Antigen synonyms:Serine/threonine-protein kinase/endoribonuclease IRE2 [Precursor]|hIRE2p|ERN2|Ire1-beta|Inositol-requiring 2|IRE1b|IRE2|Endoplasmic reticulum-to-nucleus signaling 2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: PRG2 (proteoglycan 2, bone marrow (natural killer cell activator, eosinophil granule major basic protein))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human