Anti-TSC1 Rabbit Polyclonal Antibody
Catalog # ABGEAP3470A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Tuberous sclerosis 1 protein
- Antigen symbol:TSC1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:TSC1|LAM|TSC|hamartin|MGC86987|Hamartin|KIAA0243
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: TSC1 (tuberous sclerosis 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human