Anti-NFATC2 Rabbit Polyclonal Antibody
Catalog # ABGEAP3429A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Meis homeobox 2
- Antigen symbol:MEIS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Antigen synonyms:HsT18361|Meis homeobox 2|TALE homeobox protein Meis2|Meis1|OTTHUMP00000160209|MRG1|myeloid ecotropic viral integration site 1 homolog 2|Meis homolog 2|Meis1-related gene 1|MGC2820|MEIS2
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: NFATC2 (nuclear factor of activated T-cells, cytoplasmic, calcineurin-dependent 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human