Anti-PAPSS1 Rabbit Polyclonal Antibody
ABGEAP2607B
: Abgent
- Antibody type:Primary
- Antigen name:3'-phosphoadenosine 5'-phosphosulfate Synthase 1
- Antigen symbol:PAPSS1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:C-Terminal
- Cross adsorption:No
- Antigen synonyms:ATP Sulfurylase/APS Kinase 1|Phosphoadenosine-phosphosulfate synthetase 1|SK1|ATP Sulfurylase/Adenosine 5-prime Phosphosulfate Kinase 1|ATPSK1|PAPS Synthetase 1|PAPS Synthase 1
- Sequence:A607
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: PAPSS1 (3'-phosphoadenosine 5'-phosphosulfate synthase 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human