Anti-GDF5 Rabbit Polyclonal Antibody
Catalog # ABGEAP2067A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Growth differentiation factor 5
- Antigen symbol:GDF5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal ( 1–50 aa)
- Cross adsorption:No
- Antigen synonyms:OS5|SYNS2|LAP-4|BDA1C|BMP-14|LAP4|CDMP1|SYM1B|BMP14
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: GDF5 (growth differentiation factor 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human