Anti-MTMR4 Rabbit Polyclonal Antibody
Catalog # ABGEAP17037C
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Myotubularin Related Protein 4
- Antigen symbol:MTMR4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human
- Western blot:Yes
- Epitope:Center
- Gene ID:9110
- Antigen synonyms:MGC163451|DKFZp781E194|MTMR7|Myotubularin-related protein 4|MGC163449
- Storage buffer:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Molecular weight:133.353 kDa
- Sequence:PREMKSNTSDPEIKC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0,45 mg/ml
- Shipping temperature:4 °C
- Immunogen:This MTMR4 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 695-723 amino acids from the Central region of human MTMR4.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: MTMR4 (myotubularin related protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human