Order Entry
Hungary
ContactUsLinkComponent
 
Anti-RPLP0 Rabbit Polyclonal Antibody
  ABGEAP16183A
 :  Abgent
Anti-RPLP0 Rabbit Polyclonal Antibody
  ABGEAP16183A
 :  Abgent

 

  • Antibody type:
    Primary
  • Antigen name:
    Ribosomal Protein Large P0
  • Antigen symbol:
    RPLP0
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    N-Terminal
  • Gene ID:
    6175
  • Antigen synonyms:
    P0|RPP0|60S acidic ribosomal protein P0|PRLP0|L10E|RPLP0|60S ribosomal protein L10E|LP0
  • Storage buffer:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Molecular weight:
    34.274 kDa
  • Sequence:
    REDRATWKSNYFLKC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    4 °C
  • Immunogen:
    This RPLP0 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human RPLP0.
  • Purification:
    Protein A & Peptide affinity purification
  • Pk:
    400 µl

 

 

Type: Primary
Antigen: RPLP0 (ribosomal protein, large, P0)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human