Anti-PTDSS2 Rabbit Polyclonal Antibody
Catalog # ABGEAP13051A
Supplier: Abgent
Specifications
- Antibody type:Primary
- Antigen name:Phosphatidylserine Synthase 2
- Antigen symbol:PTDSS2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Antigen synonyms:CDKN2|MLM|INK4|MTS-1|P16|P16INK4|P19|P16-INK4A|P16INK4A|INK4A|P19ARF|CDK4I|CMM2|P14ARF|P14|ARF|MTS1|TP16
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Specifications
About this item
Type: Primary
Antigen: PTDSS2 (phosphatidylserine synthase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human