Order Entry
Hungary
ContactUsLinkComponent
Anti-SUMO2 Rabbit Polyclonal Antibody
Anti-SUMO2 Rabbit Polyclonal Antibody
Catalog # ABGEAP1282A
Supplier:  Abgent
undefined
Anti-SUMO2 Rabbit Polyclonal Antibody
Catalog # ABGEAP1282A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Small ubiquitin-related modifier 2
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Epitope:
    C-Terminal
  • Cross adsorption:
    No
  • Antigen synonyms:
    Sentrin-2|SUMO-2|HSMT3|SMT3 homolog 2|Small ubiquitin-related modifier 2|Ubiquitin-like protein SMT3B|SUMO-3|SMT3B|SMT3H2
  • Storage temperature:
    Cool
  • Shipping temperature:
    Ice
  • Pk:
    400 µl

Specifications

About this item

Type: Primary
Antigen: SUMO2 (small ubiquitin-like modifier 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human