Anti-ZNF492 Rabbit Polyclonal Antibody
ABGEAP10630B
: Abgent
- Antibody type:Primary
- Antigen name:Zinc Finger Protein 492
- Antigen symbol:ZNF492
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-Terminal (1–100 aa)
- Cross adsorption:No
- Antigen synonyms:mitochondrial|Thymidine kinase 2|Mt-TK
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:400 µl
Type: Primary
Antigen: ZNF492 (zinc finger protein 492)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human