Anti-SFT2D3 Rabbit Polyclonal Antibody
Catalog # AVIVARP60656_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:SFT2 domain containing 3
- Antigen symbol:SFT2D3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:84826
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: SFT2D3 (SFT2 domain containing 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human