Anti-TCEA2 Rabbit Polyclonal Antibody
Catalog # AVIVARP58182_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Transcription Elongation Factor A (SII), 2
- Antigen symbol:TCEA2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:6919
- Antigen synonyms:TFIIS
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TCEA2 (transcription elongation factor A (SII), 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: