Anti-FBXO33 Rabbit Polyclonal Antibody
AVIVARP55943_P050
- Antibody type:Primary
- Antigen name:F-box protein 33
- Antigen symbol:FBXO33
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:middle
- Cross adsorption:none
- Gene ID:254170
- Antigen synonyms:BMND12|Fbx33|c14_5247
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: FBXO33 (F-box protein 33)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: