Anti-SETD9 Rabbit Polyclonal Antibody
AVIVARP53194_P050
- Antibody type:Primary
- Antigen name:SET domain containing 9
- Antigen symbol:SETD9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Reactivity:Human
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:133383
- Antigen synonyms:C5orf35
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: SETD9 (SET domain containing 9)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human