Anti-DCAF7 Rabbit Polyclonal Antibody
AVIVARP52110_P050
- Antibody type:Primary
- Antigen name:DDB1 and CUL4 associated factor 7
- Antigen symbol:DCAF7
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:71833
- Antigen synonyms:HAN11|2610037L01Rik|C86529|1700012F10Rik|Wdr68
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: DCAF7 (DDB1 and CUL4 associated factor 7)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: