Anti-TBCB Rabbit Polyclonal Antibody
Catalog # AVIVARP51934_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:tubulin folding cofactor B
- Antigen symbol:TBCB
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:1155
- Antigen synonyms:CKAPI|CKAP1|CG22
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TBCB (tubulin folding cofactor B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: