Anti-DGAT2 Rabbit Polyclonal Antibody
AVIVARP50084_P050
- Antibody type:Primary
- Antigen name:diacylglycerol O-acyltransferase 2
- Antigen symbol:DGAT2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:67800
- Antigen synonyms:DGAT-2|ARAT|0610010B06Rik
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Type: Primary
Antigen: DGAT2 (diacylglycerol O-acyltransferase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: