Anti-TTC6 Rabbit Polyclonal Antibody
Catalog # AVIVARP48498_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:Tetratricopeptide Repeat Domain 6
- Antigen symbol:TTC6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:115669
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TTC6 (tetratricopeptide repeat domain 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: