Anti-TPX2 Rabbit Polyclonal Antibody
Catalog # AVIVARP46176_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:TPX2, Microtubule Nucleation Factor
- Antigen symbol:TPX2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:C-terminal
- Cross adsorption:none
- Gene ID:22974
- Antigen synonyms:DIL-2|C20orf1|HCTP4|DIL2|C20orf2|p100|GD:C20orf1|FLS353|REPP86|HCA519
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: TPX2 (TPX2, microtubule-associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: