Anti-LECT1 Rabbit Polyclonal Antibody
Catalog # AVIVARP44615_P050
Supplier: Aviva Systems Biology
Specifications
- Antibody type:Primary
- Antigen name:leukocyte cell derived chemotaxin 1
- Antigen symbol:LECT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Western blot:Yes
- Epitope:N-terminal
- Cross adsorption:none
- Gene ID:11061
- Antigen synonyms:BRICD3|MYETS1|CHM1|CHM-I
- Storage temperature:Cool
- Shipping temperature:Ice
- Pk:50 µG
Specifications
About this item
Type: Primary
Antigen: LECT1 (leukocyte cell derived chemotaxin 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: