Distilling bridges, Claisen
DURAN®, borosilicate glass
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Distilling link with Liebig condenser and Claisen head. With two sockets NS 14/23 for distillation thermometer of 55 mm buli-in length. With two thread connections, GL 14 (water) and GL 18 (vacuum).
Frequently Bought Together