Order Entry
Hungary
ContactUsLinkComponent
 
 
 
 
Methanol with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
 
undefined
 
 
 
Methanol with 0,1% (v/v) trifluoroacetic acid, HiPerSolv CHROMANORM® for LC-MS
 

 

  • Nominal concentration (V/V):
    0.095 - 0.105 %
  • Evaporation residue:
    Max. 0.0001 %
  • Water:
    Max. 0.02 %
  • Ca (Calcium):
    Max. 0.2 ppm
  • K (Potassium):
    Max. 0.2 ppm
  • Mg (Magnesium):
    Max. 0.2 ppm
  • Na (Sodium):
    Max. 0.5 ppm
  • Transmittance (225 nm):
    Min. 20 %
  • Transmittance (240 nm):
    Min. 50 %
  • Transmittance (250 nm):
    Min. 80 %
  • Transmittance (260 nm):
    Min. 95 %
  • Transmittance (270 nm):
    Min. 99 %
  • Suitable for LC-MS:
    Passes test

 

Frequently Bought Together