Order Entry
Hungary
ContactUsLinkComponent
TrioPack, Acura® manual 835
TrioPack, Acura® manual 835
Catalog # 613-7254
Supplier:  Socorex Isba
undefined
TrioPack, Acura® manual 835
Catalog # 613-7254
Supplier:  Socorex Isba
Specifications
  • Description:
    TrioPack, incl. three pipettes (100 - 1000 µl / 0,5 - 5 ml / 1 - 10 ml), Qualitips® samples
  • Pk:
    1 SET
Specifications
About this item
These ready-to-use kits include three Acura® manual 835 pipettes, Qualitips® samples, QC certificates and operating instructions.