Order Entry
Hungary
ContactUsLinkComponent
Combined ecology test, AL-36B
Combined ecology test, AL-36B
Catalog # HACH180202
Supplier:  Hach
CAS Number:  
undefined
Combined ecology test, AL-36B
Catalog # HACH180202
Supplier:  Hach
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Description:
    Combined ecology test kit, AL-36B
  • Range:
    CO₂: 1,25 - 100 mg/l ; pH: 1 - 10 ; Dissolved O₂: 0,2 - 20 mg/l

Specifications

About this item

AL-36B can measure acidity, alkalinity, carbon dioxide (1,25 to 100 mg/L), dissolved oxygen (0,2 to 20 mg/L), hardness and pH (4 to 10).

  • Simple, accurate methods
  • Colour disc and drop-count titration methods

Designed to meet the portability needs of fish and wildlife conservation personnel.

Delivery: Includes twelve reagents, clippers, bottles, sample cell with cap, colour disc, viewing tubes, comparator box, measuring tube, instructions and rugged carrying case.