D-Lactose monohydrate, powder, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
Certificates
About this item
CAS Number:64044-51-5
:63-42-3
Formula:C₁₂H₂₂O₁₁·1H₂O
MDL Number:MFCD00150747
:360.32 g/mol
Melting Pt:223 °C
Storage Temperature:Freezer
Specification Test Results
- Meets ACS Reagent Chemical Requirements:≤0.005%
- For Laboratory,Research,or Manufacturing Use:≤0.03%
- Insoluble Matter:≤0.005 %
- Residue after Ignition:≤0.03 %
- Dextrose:Passes Test
- Sucrose:Passes Test
- Water (H₂O):4.0 - 6.0 %
- Dextrins, Starch:≤5 ppm
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
Specification Test Results
Frequently Bought Together