Disinfectant for instruments, Instru-med
MOLA1001000
- Description:Instru-med, disinfectant for medical instruments
- Pk:1.000 mL
Broad spectrum activity disinfectant for immersive cleaning of dental and medical instruments.
- Formulation free of aldehydes and chlorine
- Excellent cleaning and disinfecting power
- Bacrtericidal, fungicidal, sporicidal mycobactericidal and virucidal