Multi-tube racks, NovaRack
Supplier: DIVERSIFIED BIOTECH
Certificates
About this item
PP, coloured
The NovaRack™ versatile racks are suitable for test tubes and vials of different sizes on the same surface. It can accomodate 24 microcentrifuge tubes (0,5 to 2 ml), 14 tubes (12×75 mm culture tube, cryovial and a variety of blood collection tubes), 7 tubes (15 ml centrifuge tube) and one 50 ml tube. The numeric labeling for each tube cavity allows easy identification of the tubes. Stackable.
Frequently Bought Together