Order Entry
Hungary
ContactUsLinkComponent
Tissue forceps, Aesculap® SUSI®
Tissue forceps, Aesculap® SUSI®
Catalog # AESCBD557SU
Supplier:  Aesculap
Tissue forceps, Aesculap® SUSI®
Catalog # AESCBD557SU
Supplier:  Aesculap
Specifications
  • Type:
    Tissue forceps 1:2
  • Length:
    145 mm
  • Pk:
    20
Specifications
About this item
These disposable forceps are made from a high performance polymer. The special composition of this high-grade polymer ensures an outstanding quality and provides optimal mechanical properties.
  • Material ensures a safe, easy and environmentally friendly removal through incineration in the regular hospital waste removal system
  • Single use: Reduces the risk of cross-contamination
  • 1×2 teeth
  • Sterile single packed in dispenser box