Retort clamp, four pronged
Catalog # 241-7423
Supplier: BOCHEM
Specifications
- Befogóméret:0 - 120 mm
- Leírás:Clamp, four pronged
- Hosszúság:150 mm
- Anyag:Rozsdamentes acél 18/10
- Kiszerelés:1
Specifications
About this item
This four pronged retort clamp is made of stainless steel.
- Fingers are coated with silicone to protect clamped items