Fine vacuum ball valves, manual
Supplier: Welch by Gardner Denver
Certificates
About this item
These fine vacuum ball valves feature a small flange made of nickle plated aluminium.
- Nickel-plated brass casing, polished brass ball, plastic (graphitised) seals
Frequently Bought Together