Magnetic stirring bars sets, IKAFLON®
: IKA
Glass or PTFE-coated stirring bars. Used in wide range of applications. Available in 5/set or 1/set.
- Very good chemical resistance
- Beaker line bars are autoclavable
- Temperature resistant, suitable for high and low temperature applications
Frequently Bought Together