Order Entry
Hungary
ContactUsLinkComponent
 
Handheld refractometer, Rhino VET 360
  635-2542
 :  REICHERT
undefined
Handheld refractometer, Rhino VET 360
  635-2542
 :  REICHERT

 

  • Model:
    Rhino VET 360
  • Range:
    Urine: 1,001 − 1,059 SG ; Serum: 2,0 − 14,0 g/100 ml
  • Accuracy:
    Urine ±0,001 SG ; Serum ±0,2 g/100 ml
  • Description:
    Handheld refractometer
  • W×D×H:
    235×38×38 mm
  • Weight:
    162 g
  • Pk:
    1

 

 

Shock resistant refractometer for urine specific gravity and protein concentration.

  • Test takes seconds, performs without battery
  • Automatic temperature compensation (ATC) for accuracy
  • Water resistant, dustproof and sealed internal optics

The Rhino VET 360 is used for plasma, serum and peritoneal fluids of veterinary clinical applications.