IP ActivFlo Biopsy I cassettes, loose
Supplier: LEICA HISTOLOGY
Certificates
About this item
IP ActivFlo Biopsy I cassettes feature a patented 'Fluidic Flow' pore design for maximum reagent exchange. With a 0,67 mm pore size, the cassettes include an innovative V-shape pore design that pulls fluid in and forces air out. The cassettes feature sloped walls, creating a basket for small tissue management. These one piece cassettes with ‘frangible lids’ have been designed for the Leica IP C Cassette Printer, with <1% printing error rate.
- Available in loose packaging for easy printer loading
- Choose from several colours in quantities of 500
Frequently Bought Together