Pallets and sumps for drums, CW
Supplier: LACONT UMWELTTECHNIK
Certificates
About this item
These pallets and sumps made from 3 mm sheet steel are ideal for the storage of water-endangering and flammable liquids.
- Removable galvanised grid
- Ground clearance 100 mm for internal transport
- Corrosion protection by primer coating and painting or by fire-galvanised sheet steel
Frequently Bought Together