Pallets and sumps for drums, CW-60
These pallets and sumps made of 3 mm sheet steel are ideal for the storage of water-endangering and flammable liquids in containers and drums up to 60 L.
- Removable galvanised grid
- Ground clearance 100 mm for internal transport
- Corrosion protection by primer coating and painting or by fire-galvanised sheet steel
Frequently Bought Together